Learn More
Cayman Chemical Lofepramn 10mg
SDP
Catalog No. 502981943
Encompass_Preferred
A first generation tricyclic antidepressant that is extensively metabolized to desipramine; potently inhibits serotonin and norepinephrine transporters (Kds = 70 and 5.4 nM, respectively)
Catalog No. 50-298-1943 Supplier Cayman Chemical Supplier No. 2081310 MG
A first generation tricyclic antidepressant that is extensively metabolized to desipramine; potently inhibits serotonin and norepinephrine transporters (Kds = 70 and 5.4 nM, respectively)