Learn More
Medchemexpress LLC Lofexidine-d4 hydrochloride | 1206845-57-9 | 1 MG
SDP
Catalog No. 5000414018
Encompass_Preferred
Lofexidine-d4 hydrochloride (Baq-168-d4) is the deuterium labeled Lofexidine hydrochloride. Lofexidine hydrochloride is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-14018 Supplier Medchemexpress LLC Supplier No. HYB1052S1MG
Lofexidine-d4 hydrochloride (Baq-168-d4) is the deuterium labeled Lofexidine hydrochloride. Lofexidine hydrochloride is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs