Learn More
Medchemexpress LLC HY-B1052 50mg Medchemexpress, Lofexidine (hydrochloride) CAS:21498-08-8 Purity:>98%
SDP
Catalog No. 502082682
Encompass_Preferred
Medchemexpress, HY-B1052 50mg Lofexidine (hydrochloride) CAS:21498-08-8 Lofexidine (hydrochloride) is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2682 Supplier Medchemexpress LLC Supplier No. HYB105250MG
Medchemexpress, HY-B1052 50mg Lofexidine (hydrochloride) CAS:21498-08-8 Lofexidine (hydrochloride) is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.