Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lofexidine hydrochloride | 21498-08-8 | 99.7% | 100 MG
SDP

Catalog No. 5000413751
Encompass_Preferred

Lofexidine hydrochloride is a selective α2-receptor agonist primarily used in research to alleviate the physical symptoms of heroin and other types of opioid withdrawal. This product is intended for research use only and is not for patient administration.

  • Selective α2-receptor agonist
  • Aids in alleviating opioid withdrawal symptoms
  • For research use only
  • Solid appearance
  • White to off-white color
  • Molecular weight: 295.59
  • Molecular formula: C11H13Cl3N2O
  • Soluble in H2O, DMSO, and PBS

Catalog No. 50-004-13751 Supplier Medchemexpress LLC Supplier No. HYB1052100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Lofexidine hydrochloride is a selective α2-receptor agonist primarily used in research to alleviate the physical symptoms of heroin and other types of opioid withdrawal. This product is intended for research use only and is not for patient administration.

  • Selective α2-receptor agonist
  • Aids in alleviating opioid withdrawal symptoms
  • For research use only
  • Solid appearance
  • White to off-white color
  • Molecular weight: 295.59
  • Molecular formula: C11H13Cl3N2O
  • Soluble in H2O, DMSO, and PBS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.