Learn More
Selleck Chemical LLC Loganin S3835-10mg
SDP
Catalog No. 503111447
Encompass_Preferred
Loganin (Loganoside) is an iridoid glycoside first isolated from the seeds of Strychnos nux-vomica a member of the Loganiaceae family It is a non-competitive inhibitor of BACE1 with IC50 of 47 97 M an also inhibits AChE and BChE with IC50 values of 3 95 M and 33 02 M respectively
Catalog No. 50-311-1447 Supplier Selleck Chemical LLC Supplier No. S383510MG
Loganin (Loganoside) is an iridoid glycoside first isolated from the seeds of Strychnos nux-vomica a member of the Loganiaceae family It is a non-competitive inhibitor of BACE1 with IC50 of 47 97 M an also inhibits AChE and BChE with IC50 values of 3 95 M and 33 02 M respectively