Learn More
Apexbio Technology LLC Loganin 18524-94-2 20mg
SDP
Catalog No. 5000091657
Encompass_Preferred
Loganin (CAS 18524-94-2) is an iridoid glucoside that modulates key inflammatory and oxidative pathways Studies indicate that Loganin inhibits the enzymatic activity of cyclooxygenase-1 (Cox-1) and suppresses the production of tumor necrosis factor-alpha (TNF- ) implicating interference with pro-inflammatory signaling Additionally Loganin demonstrates marked free-radical-scavenging capacity and exerts inhibitory effects on melanogenesis These properties support the use of Loganin in research focused on inflammation oxidative stress and pigmentary disorders
Catalog No. 50-000-91657 Supplier Apexbio Technology LLC Supplier No. N132720
Loganin (CAS 18524-94-2) is an iridoid glucoside that modulates key inflammatory and oxidative pathways Studies indicate that Loganin inhibits the enzymatic activity of cyclooxygenase-1 (Cox-1) and suppresses the production of tumor necrosis factor-alpha (TNF- ) implicating interference with pro-inflammatory signaling Additionally Loganin demonstrates marked free-radical-scavenging capacity and exerts inhibitory effects on melanogenesis These properties support the use of Loganin in research focused on inflammation oxidative stress and pigmentary disorders