Learn More
Medchemexpress LLC Loganin | 18524-94-2 | 390.38 | 1 ML
SDP
Catalog No. 5000389190
Encompass_Preferred
Loganin is an iridoid glycoside compound with anti-inflammatory, antioxidant, and antitumor properties. It provides protective effects against acute lung injury and pulmonary fibrosis. Loganin works by upregulating the Nrf2/HO-1 signaling pathway to combat inflammation and oxidative stress, and it also reduces neuroinflammation resulting from spinal cord injury.
  • Anti-inflammatory properties
  • Antioxidant properties
  • Antitumor properties
  • Protects against acute lung injury
  • Protects against pulmonary fibrosis
  • Upregulates Nrf2/HO-1 signaling pathway
  • Combats inflammation and oxidative stress
  • Reduces neuroinflammation from spinal cord injury
Catalog No. 50-003-89190 Supplier Medchemexpress LLC Supplier No. HYN051210MM/1ML
Loganin is an iridoid glycoside compound with anti-inflammatory, antioxidant, and antitumor properties. It provides protective effects against acute lung injury and pulmonary fibrosis. Loganin works by upregulating the Nrf2/HO-1 signaling pathway to combat inflammation and oxidative stress, and it also reduces neuroinflammation resulting from spinal cord injury.
  • Anti-inflammatory properties
  • Antioxidant properties
  • Antitumor properties
  • Protects against acute lung injury
  • Protects against pulmonary fibrosis
  • Upregulates Nrf2/HO-1 signaling pathway
  • Combats inflammation and oxidative stress
  • Reduces neuroinflammation from spinal cord injury