Learn More
Medchemexpress LLC Loganin | 18524-94-2 | 390.38 | 50 MG
SDP
Catalog No. 5000390009
Encompass_Preferred
Loganin is an iridoid glycoside compound with anti-inflammatory, antioxidant, and antitumor properties. It offers protective effects against acute lung injury and pulmonary fibrosis. Loganin protects against LPS-mediated inflammation and oxidative stress by upregulating the Nrf2/HO-1 signaling pathway, and it reduces neuroinflammation caused by spinal cord injury (SCI).
  • Anti-inflammatory, antioxidant, and antitumor properties
  • Protects against acute lung injury and pulmonary fibrosis
  • Upregulates nrf2/HO-1 signaling pathway
  • Reduces neuroinflammation caused by spinal cord injury
  • High purity
  • White to off-white solid appearance
Catalog No. 50-003-90009 Supplier Medchemexpress LLC Supplier No. HYN051250MG
Loganin is an iridoid glycoside compound with anti-inflammatory, antioxidant, and antitumor properties. It offers protective effects against acute lung injury and pulmonary fibrosis. Loganin protects against LPS-mediated inflammation and oxidative stress by upregulating the Nrf2/HO-1 signaling pathway, and it reduces neuroinflammation caused by spinal cord injury (SCI).
  • Anti-inflammatory, antioxidant, and antitumor properties
  • Protects against acute lung injury and pulmonary fibrosis
  • Upregulates nrf2/HO-1 signaling pathway
  • Reduces neuroinflammation caused by spinal cord injury
  • High purity
  • White to off-white solid appearance