Learn More
TARGETMOL CHEMICALS INC LOGANIN 25MG
SDP
Catalog No. 502577050
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Loganin (Loganoside) an iridoid glucoside has been reported to be an inhibitor of Cox-1 and also noted to suppress TNF-(alpha) formation. This compound also has displayed potent free-radical-scavenging activity and inhibitory effects on melanogenesis. purity: 99%
Catalog No. 50-257-7050 Supplier TARGETMOL CHEMICALS INC Supplier No. T275925MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Loganin (Loganoside) an iridoid glucoside has been reported to be an inhibitor of Cox-1 and also noted to suppress TNF-(alpha) formation. This compound also has displayed potent free-radical-scavenging activity and inhibitory effects on melanogenesis. purity: 99%