Learn More
Cayman Chemical Loganic Acid, Purity: ≥98%, CAS: 22255-40-9, 10 mg
SDP
Catalog No. 502937297
Encompass_Preferred
An iridoid glycoside; stimulates GLP-1 secretion by NCI H716 human enteroendocrine colorectal cancer cells at 1 μM; inhibits dexamethasone-, 3-isobutyl-1-methylxanthine-, and insulin-induced differentiation of mouse 3T3-L1 fibroblasts into adipocytes at 10 μg/ml; decreases the plasma atherogenic index and cardiac risk ratio, as well as plasma levels of IL-6, TNF-α, and oxLDL in a cholesterol-rich diet-induced rabbit model of atherosclerosis
Catalog No. 50-293-7297 Supplier Cayman Chemical Supplier No. 2840210 MG
An iridoid glycoside; stimulates GLP-1 secretion by NCI H716 human enteroendocrine colorectal cancer cells at 1 μM; inhibits dexamethasone-, 3-isobutyl-1-methylxanthine-, and insulin-induced differentiation of mouse 3T3-L1 fibroblasts into adipocytes at 10 μg/ml; decreases the plasma atherogenic index and cardiac risk ratio, as well as plasma levels of IL-6, TNF-α, and oxLDL in a cholesterol-rich diet-induced rabbit model of atherosclerosis