Learn More
Aviva Systems Biology Corp LOH11CR2A Antibody - N-terminal region (ARP55093_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55093_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOH11CR2A. It was validated on Western Blot. VWA5A,von Willebrand factor A domain containing 5A,BCSC1; BCSC-1; LOH11CR2A; VWA5A,von Willebrand factor A domain-containing protein 5A,von Willebrand factor A domain-containing protein 5A,ortholog of mouse AW551984,breast cancer suppressor candidate 1,loss of heterozygosity, 11, chromosomal region 2, gene A,loss of heterozygosity 11 chromosomal region 2 gene A proteinThe immunogen is a synthetic peptide directed towards the N terminal region of human LOH11CR2A. Alternate names: BCSC-1, BCSC1, LOH11CR2AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-001