Learn More
Carnabio Usa Inc LOK(STK10)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 071155
Encompass
LOK(STK10), Full-length, Wild type, Amino Acid 1-968, BAA35073.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0486