Learn More
Hardy Diagnostics LokTight™ Sabouraud Dextrose Agar (SabDex) with Lecithin and Tween, Contact Plate, USP, Irradiated, Triple Bagged
Shop All Hardy Diagnostics Products

Description
- Recommended for the detection and enumeration of fungi (yeast and mold) from environmental sources in a clean room setting. It is a triple bagged contact plate, also commonly referred to as Rodac™, for surface monitoring. Gamma irradiated for sterility assurance
- Three Easy Steps:
- Remove lid
- Gently press plate to surface without twisting or sliding
- Twist clockwise until frosted bands line up to lock lid into place
- Features and Benefits:
- Triple bagged: Transparent PET outer and middle bag for distortion-free viewing, breathable transparent inner bag
- The LokTight™ locking lid is an optional feature designed to keep lids securely in place after sampling and throughout transport to your lab
- Each plate has a grid molded into the bottom of the plate to aid in the counting of microorganisms
- A Certificate of Analysis is available online for each lot
- Room temperature storage-sterile by irradiation
- Validated for a Sterility Assurance Level (SAL) of 10⁻⁶
Specifications
Specifications
| Form | Agar |
| Manufacturing Origin | US with Globally Sourced Materials |
| Product Type | Agar Plate, Contact Plate |
| Sterility | Sterile |
| Target Organism Class | Fungi, Yeast, Molds |
| Test Type | Microbial Detection, Media Testing, Environmental Test, Growth Promotion Testing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.