Learn More
Medchemexpress LLC HY-101035 100mg Medchemexpress, LOM612 CAS:2173232-79-4 Purity:>98%
SDP
Catalog No. 501932016
Encompass_Preferred
Medchemexpress, HY-101035 100mg LOM612 CAS:2173232-79-4 LOM612 is a potent activator of FOXO nuclear translocation, with an EC 50 value of 1.5 μM in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2016 Supplier Medchemexpress LLC Supplier No. HY101035100MG
Medchemexpress, HY-101035 100mg LOM612 CAS:2173232-79-4 LOM612 is a potent activator of FOXO nuclear translocation, with an EC 50 value of 1.5 μM in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.