Learn More
Medchemexpress LLC Lomefloxacin-d5 (hydrochloride) | 98079-52-8 | 99.9% | 392.84 | C17H15D5ClF2N3O3 | 10 MG
SDP
Catalog No. 5000297359
Encompass_Preferred
Lomefloxacin-d5 (hydrochloride) is the deuterium-labeled analogue of the antibiotic lomefloxacin, supplied as a hydrochloride salt and intended for use as an analytical reference standard and tracer for quantitative methods. It is formulated and tested for high purity and supplied with supporting COA and SDS documentation.
  • Deuterated internal standard for quantitative NMR, GC-MS, and LC-MS analysis.
  • High reported purity by LC-MS (≈99.9%).
  • Provided as a hydrochloride salt for improved stability.
  • Supplied with certificate of analysis and SDS for traceability.
  • Recommended storage at 4°C; in solvent, -80°C for long-term storage.
Catalog No. 50-002-97359 Supplier Medchemexpress LLC Supplier No. HYB0455S10MG
Add to Cart
Add to Cart
Lomefloxacin-d5 (hydrochloride) is the deuterium-labeled analogue of the antibiotic lomefloxacin, supplied as a hydrochloride salt and intended for use as an analytical reference standard and tracer for quantitative methods. It is formulated and tested for high purity and supplied with supporting COA and SDS documentation.
  • Deuterated internal standard for quantitative NMR, GC-MS, and LC-MS analysis.
  • High reported purity by LC-MS (≈99.9%).
  • Provided as a hydrochloride salt for improved stability.
  • Supplied with certificate of analysis and SDS for traceability.
  • Recommended storage at 4°C; in solvent, -80°C for long-term storage.