Learn More
Medchemexpress LLC HY-B0455 100mg Medchemexpress, Lomefloxacin (hydrochloride) CAS:98079-52-8 Purity:>98%
SDP
Catalog No. 502082546
Encompass_Preferred
Medchemexpress, HY-B0455 100mg Lomefloxacin (hydrochloride) CAS:98079-52-8 Lomefloxacin hydrochloride is a broad-spectrum quinolone antibiotic, with antimicrobial activity. Lomefloxacin hydrochloride is used for the research of bronchitis, urinary tract infection, conjunctivitis, otitis externa, and otitis media. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2546 Supplier Medchemexpress LLC Supplier No. HYB0455100MG
Medchemexpress, HY-B0455 100mg Lomefloxacin (hydrochloride) CAS:98079-52-8 Lomefloxacin hydrochloride is a broad-spectrum quinolone antibiotic, with antimicrobial activity. Lomefloxacin hydrochloride is used for the research of bronchitis, urinary tract infection, conjunctivitis, otitis externa, and otitis media. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.