Learn More
Medchemexpress LLC Lomefloxacin (hydrochloride) | 98079-52-8 | 100.0% | 1 G
SDP
Catalog No. 5000408469
Encompass_Preferred
Lomefloxacin hydrochloride (NY-198 hydrochloride) is an orally active difluoroquinolone antibiotic that prevents DNA supercoiling and replication by inhibiting bacterial topoisomerase II. It induces ROS production and Apoptosis, and exhibits broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria. Lomefloxacin hydrochloride also demonstrates anticancer effects against melanoma. It can be used in the study of systemic bacterial infections (such as Salmonella typhimurium infections), skin, and melanoma.
  • Orally active difluoroquinolone antibiotic
  • Inhibits bacterial topoisomerase II to prevent DNA supercoiling and replication
  • Induces ROS production and apoptosis
  • Exhibits broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria
  • Demonstrates anticancer effects against melanoma
  • Used in the study of systemic bacterial infections, skin, and melanoma
Catalog No. 50-004-08469 Supplier Medchemexpress LLC Supplier No. HYB04551G
Lomefloxacin hydrochloride (NY-198 hydrochloride) is an orally active difluoroquinolone antibiotic that prevents DNA supercoiling and replication by inhibiting bacterial topoisomerase II. It induces ROS production and Apoptosis, and exhibits broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria. Lomefloxacin hydrochloride also demonstrates anticancer effects against melanoma. It can be used in the study of systemic bacterial infections (such as Salmonella typhimurium infections), skin, and melanoma.
  • Orally active difluoroquinolone antibiotic
  • Inhibits bacterial topoisomerase II to prevent DNA supercoiling and replication
  • Induces ROS production and apoptosis
  • Exhibits broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria
  • Demonstrates anticancer effects against melanoma
  • Used in the study of systemic bacterial infections, skin, and melanoma