Learn More
Medchemexpress LLC Lomefloxacin hydrochloride | 98079-52-8 | 99.97% | 5 G
SDP
Catalog No. 5000406328
Encompass_Preferred
Lomefloxacin hydrochloride (NY-198 hydrochloride) is an orally active difluoroquinolone antibiotic. It prevents DNA supercoiling and replication by inhibiting bacterial topoisomerase II. This compound induces ROS production and apoptosis, demonstrating broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria, and also exhibits anticancer effects against melanoma.
  • Orally active difluoroquinolone antibiotic
  • Inhibits bacterial topoisomerase II
  • Induces ROS production and apoptosis
  • Broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria
  • Exhibits anticancer effects against melanoma
  • Useful in studying systemic bacterial infections
  • Reduces viable cell counts of common bacteria
  • Decreases viability and induces ROS production in melanoma cells
Catalog No. 50-004-06328 Supplier Medchemexpress LLC Supplier No. HYB04555G
Lomefloxacin hydrochloride (NY-198 hydrochloride) is an orally active difluoroquinolone antibiotic. It prevents DNA supercoiling and replication by inhibiting bacterial topoisomerase II. This compound induces ROS production and apoptosis, demonstrating broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria, and also exhibits anticancer effects against melanoma.
  • Orally active difluoroquinolone antibiotic
  • Inhibits bacterial topoisomerase II
  • Induces ROS production and apoptosis
  • Broad-spectrum bactericidal activity against Gram-positive and Gram-negative bacteria
  • Exhibits anticancer effects against melanoma
  • Useful in studying systemic bacterial infections
  • Reduces viable cell counts of common bacteria
  • Decreases viability and induces ROS production in melanoma cells