Learn More
Cayman Chemical Lomefloxacin (hydrochloride), Purity: ≥98%, CAS: 98079-52-8, 10 g
SDP
Catalog No. 502939963
Encompass_Preferred
A broad-spectrum fluoroquinolone antibiotic; inhibits growth of N. gonorrhoeae, E. coli, K. pneumoniae, P. vulgaris, S. epidermidis, and M. morganii (MIC90s = 0.1, 0.39, 0.39, 0.39, 0.39, and 0.78 μg/ml, respectively); inhibits growth of S. aureus and MRSA (MIC90 = 3.13 μg/ml for both) and of H. influenzae and ampicillin-resistant H. influenzae (MIC90 = 0.1 μg/ml for both); inhibits the growth of P. mirabilis, E. coli, K. pneumoniae, and S. aureus (ED50s = 1.39, 1.45, 1.78, and 6.66 mg/kg, respectively) in mouse models of systemic infection
Catalog No. 50-293-9963 Supplier Cayman Chemical Supplier No. 2827110 G
A broad-spectrum fluoroquinolone antibiotic; inhibits growth of N. gonorrhoeae, E. coli, K. pneumoniae, P. vulgaris, S. epidermidis, and M. morganii (MIC90s = 0.1, 0.39, 0.39, 0.39, 0.39, and 0.78 μg/ml, respectively); inhibits growth of S. aureus and MRSA (MIC90 = 3.13 μg/ml for both) and of H. influenzae and ampicillin-resistant H. influenzae (MIC90 = 0.1 μg/ml for both); inhibits the growth of P. mirabilis, E. coli, K. pneumoniae, and S. aureus (ED50s = 1.39, 1.45, 1.78, and 6.66 mg/kg, respectively) in mouse models of systemic infection