Learn More
Cayman Chemical LomeguatrIb 10mg
SDP
Catalog No. 502952110
Encompass_Preferred
A modified quanine base which acts as a pseudosubstrate inactivator of MGMT (IC50 = ~3 nM)
Catalog No. 50-295-2110 Supplier Cayman Chemical Supplier No. 1173210 MG
A modified quanine base which acts as a pseudosubstrate inactivator of MGMT (IC50 = ~3 nM)