Learn More
Medchemexpress LLC Lomeguatrib | 192441-08-0 | 99.8% | 326.17 | 100 MG
SDP
Catalog No. 5000348458
Encompass_Preferred
Lomeguatrib is an O6-methylguanine-DNA methyltransferase (MGMT) inhibitor. It exhibits an IC50 of 9 nM in cell-free assays and 6 nM in MCF-7 cells.
  • Acts as an O6-methylguanine-DNA methyltransferase (MGMT) inhibitor.
  • Shows an IC50 of 9 nM in cell-free assays.
  • Exhibits an IC50 of 6 nM in MCF-7 cells.
  • Increases the growth inhibitory effects of temozolomide in MCF-7 cells.
  • Completely inactivates MGMT within 2 hours at 20 mg/kg i.p.
Catalog No. 50-003-48458 Supplier Medchemexpress LLC Supplier No. HY13668100MG
Lomeguatrib is an O6-methylguanine-DNA methyltransferase (MGMT) inhibitor. It exhibits an IC50 of 9 nM in cell-free assays and 6 nM in MCF-7 cells.
  • Acts as an O6-methylguanine-DNA methyltransferase (MGMT) inhibitor.
  • Shows an IC50 of 9 nM in cell-free assays.
  • Exhibits an IC50 of 6 nM in MCF-7 cells.
  • Increases the growth inhibitory effects of temozolomide in MCF-7 cells.
  • Completely inactivates MGMT within 2 hours at 20 mg/kg i.p.