Learn More
Cayman Chemical LOMEGUATRIB 50MG
SDP
Catalog No. 502952111
Encompass_Preferred
A modified quanine base which acts as a pseudosubstrate inactivator of MGMT (IC50 = ~3 nM)
Catalog No. 50-295-2111 Supplier Cayman Chemical Supplier No. 1173250 MG
A modified quanine base which acts as a pseudosubstrate inactivator of MGMT (IC50 = ~3 nM)