Learn More
TARGETMOL CHEMICALS INC LOMEGUATRIB 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T249550MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lomeguatrib (PaTrin-2) a modified guanine base inhibits the activity of DNA repair protein O(6)-alkylguanine-DNA alkyltransferase (MGMT) . purity: 99%
Catalog No. 50-258-0774