Learn More
Abcam Lomeguatrib, MGMT inhibitor, 5MG
Catalog No. 502706194
Encompass_Preferred
MW 326.17 Da, Purity >98%. Potent O⁶-methylguanine-DNA methyltransferase (MGMT) inhibitor (IC₅₀ = 9 nM). Inhibits recombinant human ATase (IC₅₀ = 5 nM). Shows antitumor effects in vivo. Enhances antitumor effects of temozolomide (ab141055).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6194 Supplier Abcam Supplier No. AB1427415MG
MW 326.17 Da, Purity >98%. Potent O⁶-methylguanine-DNA methyltransferase (MGMT) inhibitor (IC₅₀ = 9 nM). Inhibits recombinant human ATase (IC₅₀ = 5 nM). Shows antitumor effects in vivo. Enhances antitumor effects of temozolomide (ab141055).
The product is subject to the following: Abcam Restricted Use Statement