Learn More
Selleck Chemical LLC Lomerizine 2HCl S4084-1g
SDP
Catalog No. 503108723
Encompass_Preferred
Lomerizine dihydrochloride (KB-2796) is a relatively new L- and T-type calcium channel blocker used in the treatment of migraine
Catalog No. 50-310-8723 Supplier Selleck Chemical LLC Supplier No. S40841G
Add to Cart
Add to Cart
Lomerizine dihydrochloride (KB-2796) is a relatively new L- and T-type calcium channel blocker used in the treatment of migraine