Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lomerizine dihydrochloride | 101477-54-7 | 100.0% | 541.46 | C27H32Cl2F2N2O3 | 10 MG
SDP

Catalog No. 5000297623
Encompass_Preferred

Lomerizine dihydrochloride is a research chemical that acts as an antagonist of L-type and T-type voltage-gated calcium channels. It is used in preclinical and in vitro studies investigating calcium signaling, migraine mechanisms, and vascular effects.

  • Antagonist of L- and T-type voltage-gated calcium channels.
  • High reported purity (99.95%).
  • Molecular weight 541.46 g/mol.
  • CAS number 101477-54-7 for unambiguous identification.
  • Supplied as a solid in 10 mg quantities for small-scale research use.
  • Intended for research purposes; follow safety and handling guidance.

Catalog No. 50-002-97623 Supplier Medchemexpress LLC Supplier No. HYB0768A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lomerizine dihydrochloride is a research chemical that acts as an antagonist of L-type and T-type voltage-gated calcium channels. It is used in preclinical and in vitro studies investigating calcium signaling, migraine mechanisms, and vascular effects.

  • Antagonist of L- and T-type voltage-gated calcium channels.
  • High reported purity (99.95%).
  • Molecular weight 541.46 g/mol.
  • CAS number 101477-54-7 for unambiguous identification.
  • Supplied as a solid in 10 mg quantities for small-scale research use.
  • Intended for research purposes; follow safety and handling guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.