Learn More
Medchemexpress LLC Lomerizine dihydrochloride | 101477-54-7 | 99.9% | 100 MG
SDP
Catalog No. 5000409193
Encompass_Preferred
Lomerizine dihydrochloride is an antagonist of L- and T-type voltage-gated calcium channels. It is intended for research use only.
  • Antagonist of L- and T-type voltage-gated calcium channels
  • Neuroprotective in motor neurons
  • Increases cerebral blood flow
  • Reverses multidrug resistance by inhibiting P-gp function
Catalog No. 50-004-09193 Supplier Medchemexpress LLC Supplier No. HYB0768A100MG
Lomerizine dihydrochloride is an antagonist of L- and T-type voltage-gated calcium channels. It is intended for research use only.
  • Antagonist of L- and T-type voltage-gated calcium channels
  • Neuroprotective in motor neurons
  • Increases cerebral blood flow
  • Reverses multidrug resistance by inhibiting P-gp function