Learn More
Medchemexpress LLC Lomerizine dihydrochloride | 101477-54-7 | 99.95% | 1 ML
SDP
Catalog No. 5000408039
Encompass_Preferred
Lomerizine dihydrochloride, also known as KB-2796, is a potent antagonist of L- and T-type voltage-gated calcium channels. This chemical is supplied at a concentration of 10mM in 1mL of DMSO. It is crucial for researchers studying calcium channel modulation and related neurological processes.
  • Antagonist of L-type voltage-gated calcium channels
  • Antagonist of T-type voltage-gated calcium channels
  • Packaged as a 10mM solution in DMSO
  • Recommended storage at 4°C, sealed, away from moisture
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
  • Suitable for shipping at room temperature for durations less than 2 weeks
Catalog No. 50-004-08039 Supplier Medchemexpress LLC Supplier No. HYB0768A10MM/1ML
Lomerizine dihydrochloride, also known as KB-2796, is a potent antagonist of L- and T-type voltage-gated calcium channels. This chemical is supplied at a concentration of 10mM in 1mL of DMSO. It is crucial for researchers studying calcium channel modulation and related neurological processes.
  • Antagonist of L-type voltage-gated calcium channels
  • Antagonist of T-type voltage-gated calcium channels
  • Packaged as a 10mM solution in DMSO
  • Recommended storage at 4°C, sealed, away from moisture
  • Stable in solvent for 6 months at -80°C or 1 month at -20°C
  • Suitable for shipping at room temperature for durations less than 2 weeks