Learn More
Medchemexpress LLC Lomerizine dihydrochloride | 101477-54-7 | 99.95% | 50 MG
SDP
Catalog No. 5000408866
Encompass_Preferred
Lomerizine dihydrochloride is an antagonist of L- and T-type voltage-gated calcium channels. It also antagonizes transient receptor potential channel 5. Lomerizine is a dual L/T-type channel blocker that is used for prophylaxis of migraine.
  • Antagonist of L- and T-type voltage-gated calcium channels.
  • Demonstrates neuroprotective effects.
  • Possesses activity in reversing multidrug resistance.
  • Increases cerebral blood flow.
Catalog No. 50-004-08866 Supplier Medchemexpress LLC Supplier No. HYB0768A50MG
Lomerizine dihydrochloride is an antagonist of L- and T-type voltage-gated calcium channels. It also antagonizes transient receptor potential channel 5. Lomerizine is a dual L/T-type channel blocker that is used for prophylaxis of migraine.
  • Antagonist of L- and T-type voltage-gated calcium channels.
  • Demonstrates neuroprotective effects.
  • Possesses activity in reversing multidrug resistance.
  • Increases cerebral blood flow.