Learn More
Apexbio Technology LLC Lomerizine HCl 101477-54-7 50mg
SDP
Catalog No. 502240322
Encompass_Preferred
Lomerizine HCl (CAS 101477-54-7) is a small molecule antagonist of voltage-gated calcium channels exhibiting inhibitory activity on both L-type and T-type subtypes By modulating calcium influx across neuronal membranes lomerizine HCl is proposed to attenuate neuronal excitability associated with migraine pathophysiology This compound is utilized in biomedical research to investigate calcium channel function and the molecular mechanisms underlying migraine and related neurological disorders
This item is not returnable. View return policy
Catalog No. 50-224-0322 Supplier Apexbio Technology LLC Supplier No. B178250
Lomerizine HCl (CAS 101477-54-7) is a small molecule antagonist of voltage-gated calcium channels exhibiting inhibitory activity on both L-type and T-type subtypes By modulating calcium influx across neuronal membranes lomerizine HCl is proposed to attenuate neuronal excitability associated with migraine pathophysiology This compound is utilized in biomedical research to investigate calcium channel function and the molecular mechanisms underlying migraine and related neurological disorders