Learn More
Medchemexpress LLC Lometrexol | 106400-81-1 | MFCD00866319 | >98.0% | C21H25N5O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1452110MG
Encompass_Preferred
Lometrexol is an antipurine antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT) and human serine hydroxymethyltransferase 1/2 (hSHMT1/2). It is used in biochemical and oncology research to study de novo purine synthesis, cell proliferation, and apoptosis.
  • Potent inhibitor of GARFT and hSHMT1/2 for mechanistic studies.
  • Suitable for in vitro biochemical and cellular assays.
  • Available in small research pack sizes for screening and formulation.
  • Well characterized with reported molecular formula C21H25N5O6.
  • Supplied with supporting product documentation and datasheet for characterization.
Catalog No. 50-002-60514