Learn More
Medchemexpress LLC Lometrexol | 106400-81-1 | MFCD00866319 | 99.0% | 443.45 g·mol⁻¹ | C21H25N5O6 | 5 MG
SDP
Catalog No. 5000221209
Encompass_Preferred
Lometrexol (CAS 106400-81-1) is an antipurine antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT). It is used in biochemical and cellular studies to probe de novo purine biosynthesis, assess effects on cell proliferation and apoptosis, and characterize enzyme inhibition.
  • Inhibits glycinamide ribonucleotide formyltransferase (GARFT).
  • Suitable for biochemical and cell-based assays.
  • Available in mg-scale research pack sizes.
  • High purity for research applications.
  • Useful for studies of purine biosynthesis and cytotoxicity.
Catalog No. 50-002-21209 Supplier Medchemexpress LLC Supplier No. HY145215MG
Lometrexol (CAS 106400-81-1) is an antipurine antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT). It is used in biochemical and cellular studies to probe de novo purine biosynthesis, assess effects on cell proliferation and apoptosis, and characterize enzyme inhibition.
  • Inhibits glycinamide ribonucleotide formyltransferase (GARFT).
  • Suitable for biochemical and cell-based assays.
  • Available in mg-scale research pack sizes.
  • High purity for research applications.
  • Useful for studies of purine biosynthesis and cytotoxicity.