Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lometrexol disodium | 120408-07-3 | 98.0% | 487.42 | C21H23N5Na2O6 | 5 MG
SDP

Catalog No. 5000221113
Encompass_Preferred

Lometrexol disodium is an antipurine antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT) and serine hydroxymethyltransferase 1/2 (hSHMT1/2), disrupting de novo purine synthesis and leading to cell cycle arrest and apoptosis. It is supplied as a disodium salt for biochemical and cellular studies.

  • Potent GARFT and hSHMT1/2 inhibitor used to study purine metabolism.
  • High purity: 98.0% suitable for analytical and biological assays.
  • Molecular formula C21H23N5Na2O6 and molecular weight 487.42.
  • Provided as a solid powder in small research pack sizes for convenience.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-21113 Supplier Medchemexpress LLC Supplier No. HY14521A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lometrexol disodium is an antipurine antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT) and serine hydroxymethyltransferase 1/2 (hSHMT1/2), disrupting de novo purine synthesis and leading to cell cycle arrest and apoptosis. It is supplied as a disodium salt for biochemical and cellular studies.

  • Potent GARFT and hSHMT1/2 inhibitor used to study purine metabolism.
  • High purity: 98.0% suitable for analytical and biological assays.
  • Molecular formula C21H23N5Na2O6 and molecular weight 487.42.
  • Provided as a solid powder in small research pack sizes for convenience.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.