Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lometrexol disodium | 120408-07-3 | MFCD00239212 | 98.0% | C21H23N5Na2O6 | 10MG
SDP

Catalog No. 5000261150
Encompass_Preferred

Lometrexol disodium is an antifolate research reagent that inhibits glycinamide ribonucleotide formyltransferase (GARFT), disrupting de novo purine biosynthesis. Supplied as the disodium salt, it is intended for laboratory research use.

  • Purity 98.0%.
  • Chemical formula C21H23N5Na2O6.
  • Molecular weight 487.42 g/mol.
  • Appearance solid, white to light yellow.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Research applications include study of purine biosynthesis inhibition.

Catalog No. 50-002-61150 Supplier Medchemexpress LLC Supplier No. HY14521A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lometrexol disodium is an antifolate research reagent that inhibits glycinamide ribonucleotide formyltransferase (GARFT), disrupting de novo purine biosynthesis. Supplied as the disodium salt, it is intended for laboratory research use.

  • Purity 98.0%.
  • Chemical formula C21H23N5Na2O6.
  • Molecular weight 487.42 g/mol.
  • Appearance solid, white to light yellow.
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 50 mg, 100 mg.
  • Research applications include study of purine biosynthesis inhibition.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.