Learn More
Medchemexpress LLC Lometrexol hydrate | 1435784-14-7 | 99.0% | 461.47 g/mol | C21H27N5O7 | 5 MG
SDP
Catalog No. 5000221452
Encompass_Preferred
Lometrexol hydrate is an antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT), disrupting de novo purine synthesis and producing antiproliferative effects in biochemical and cell-based studies. It is supplied as a solid suitable for laboratory assays and preclinical research.
  • Inhibits GARFT to block de novo purine synthesis.
  • Suitable for in vitro biochemical and cell biology studies.
  • High purity for reproducible experimental results.
  • Stable crystalline solid, convenient for handling and storage.
  • Supplied in research-scale quantities for assay development.
Catalog No. 50-002-21452 Supplier Medchemexpress LLC Supplier No. HY14521B5MG
Lometrexol hydrate is an antifolate research compound that inhibits glycinamide ribonucleotide formyltransferase (GARFT), disrupting de novo purine synthesis and producing antiproliferative effects in biochemical and cell-based studies. It is supplied as a solid suitable for laboratory assays and preclinical research.
  • Inhibits GARFT to block de novo purine synthesis.
  • Suitable for in vitro biochemical and cell biology studies.
  • High purity for reproducible experimental results.
  • Stable crystalline solid, convenient for handling and storage.
  • Supplied in research-scale quantities for assay development.