Learn More
Selleck Chemical LLC Lomibuvir (VX-222) S1480-5mg
SDP
Catalog No. 503110964
Encompass_Preferred
Lomibuvir (VX-222 VCH-222) is a novel potent and selective inhibitor of HCV polymerase with IC50 of 0 94-1 2 M 15 3-fold less effective for mutant M423T and 108-fold less effective for mutant I482L Phase 2
Catalog No. 50-311-0964 Supplier Selleck Chemical LLC Supplier No. S14805MG
Lomibuvir (VX-222 VCH-222) is a novel potent and selective inhibitor of HCV polymerase with IC50 of 0 94-1 2 M 15 3-fold less effective for mutant M423T and 108-fold less effective for mutant I482L Phase 2