Learn More
Medchemexpress LLC Lomitapide | 182431-12-5 | 99.6% | 693.72 | 25 MG
SDP
Catalog No. 5000363286
Encompass_Preferred
Lomitapide is a potent inhibitor of microsomal triglyceride-transfer protein (MTP), demonstrating an IC50 of 8 nM in vitro.
  • Inhibits microsomal triglyceride-transfer protein (MTP) with an IC50 of 8 nM in vitro.
  • Reduces plasma concentrations of low density lipoprotein cholesterol (LDL-C) and triglycerides.
  • Indicated for the treatment of Homozygous Familial Hypercholesterolemia (HoFH).
  • Metabolized via cytochrome P-450 (CYP) isoenzyme 3A4.
  • Bioavailability of 7.1% and a mean half-life of 39.7 hours for the 50-mg capsule.
  • Exhibits antiproliferative activity against human HepG2 cells.
  • Appears as a solid, white to off-white substance.
Catalog No. 50-003-63286 Supplier Medchemexpress LLC Supplier No. 25MGHY14667
Lomitapide is a potent inhibitor of microsomal triglyceride-transfer protein (MTP), demonstrating an IC50 of 8 nM in vitro.
  • Inhibits microsomal triglyceride-transfer protein (MTP) with an IC50 of 8 nM in vitro.
  • Reduces plasma concentrations of low density lipoprotein cholesterol (LDL-C) and triglycerides.
  • Indicated for the treatment of Homozygous Familial Hypercholesterolemia (HoFH).
  • Metabolized via cytochrome P-450 (CYP) isoenzyme 3A4.
  • Bioavailability of 7.1% and a mean half-life of 39.7 hours for the 50-mg capsule.
  • Exhibits antiproliferative activity against human HepG2 cells.
  • Appears as a solid, white to off-white substance.