Learn More
Medchemexpress LLC Lomitapide mesylate | 202914-84-9 | 99.6% | 789.83 | 25 MG
SDP
Catalog No. 5000363460
Encompass_Preferred
Lomitapide mesylate is a highly effective microsomal triglyceride transfer protein (MTP) inhibitor, demonstrating an in vitro IC50 value of 8 nM. It inhibits ApoB lipoprotein secretion in human HepG2 cells with an ED50 of 0.8 nM. This compound is provided as a solid, appearing white to off-white, and is suitable for research applications.
  • Highly effective MTP protein inhibitor
  • In vitro IC50 value of 8 nM
  • Inhibits ApoB lipoprotein secretion in HepG2 cells
  • Solubility in DMSO: 100 mg/mL
  • Solid appearance, white to off-white
  • For research use only
Catalog No. 50-003-63460 Supplier Medchemexpress LLC Supplier No. 25MGHY14668
Lomitapide mesylate is a highly effective microsomal triglyceride transfer protein (MTP) inhibitor, demonstrating an in vitro IC50 value of 8 nM. It inhibits ApoB lipoprotein secretion in human HepG2 cells with an ED50 of 0.8 nM. This compound is provided as a solid, appearing white to off-white, and is suitable for research applications.
  • Highly effective MTP protein inhibitor
  • In vitro IC50 value of 8 nM
  • Inhibits ApoB lipoprotein secretion in HepG2 cells
  • Solubility in DMSO: 100 mg/mL
  • Solid appearance, white to off-white
  • For research use only