Learn More
Medchemexpress LLC Lomitapide mesylate | 202914-84-9 | MFCD19443682 | 99.6% | C40H41F6N3O5S | 10MG
SDP
Catalog No. 5000259563
Encompass_Preferred
Lomitapide mesylate is the methanesulfonate salt of lomitapide, a potent inhibitor of microsomal triglyceride-transfer protein (MTP) used as a research reagent. It has a reported in vitro IC50 of 8 nM and is supplied as a high-purity solid suitable for biochemical assays, cell-based studies, and formulation for animal dosing.
  • Has potent in vitro activity (IC50 8 nM) against MTP.
  • Supplied at high purity (≈99.6%).
  • Soluble in DMSO; manufacturer provides recommended in vivo vehicle formulations.
  • Available in multiple pack sizes from milligram to gram scale.
  • Suitable for biochemical assays, cell-based assays, and in vivo formulation work.
Catalog No. 50-002-59563 Supplier Medchemexpress LLC Supplier No. HY1466810MG
Lomitapide mesylate is the methanesulfonate salt of lomitapide, a potent inhibitor of microsomal triglyceride-transfer protein (MTP) used as a research reagent. It has a reported in vitro IC50 of 8 nM and is supplied as a high-purity solid suitable for biochemical assays, cell-based studies, and formulation for animal dosing.
  • Has potent in vitro activity (IC50 8 nM) against MTP.
  • Supplied at high purity (≈99.6%).
  • Soluble in DMSO; manufacturer provides recommended in vivo vehicle formulations.
  • Available in multiple pack sizes from milligram to gram scale.
  • Suitable for biochemical assays, cell-based assays, and in vivo formulation work.