Learn More
TARGETMOL CHEMICALS INC Lomitapide Mesylate 25MG
SDP
Catalog No. 502877741
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Lomitapide Mesylate (BMS-201038 mesylate) is a methanesulfonic acid form of lomitapide, a small molecule inhibitor of microsomal triglyceride transfer protein. Purity 99.65%
Catalog No. 50-287-7741 Supplier TARGETMOL CHEMICALS INC Supplier No. T254525MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Lomitapide Mesylate (BMS-201038 mesylate) is a methanesulfonic acid form of lomitapide, a small molecule inhibitor of microsomal triglyceride transfer protein. Purity 99.65%