Learn More
Selleck Chemical LLC Lomitapide Mesylate
SDP
Catalog No. 5000000958
Encompass_Preferred
Lomitapide Mesylate(AEGR-733 Mesylate BMS-2138 Mesylate) is a potent microsomal triglyceride transfer protein (MTP) inhibitor used in the treatment of familial hypercholesterolemia
Catalog No. 50-000-00958 Supplier Selleck Chemical LLC Supplier No. S763350MG
Lomitapide Mesylate(AEGR-733 Mesylate BMS-2138 Mesylate) is a potent microsomal triglyceride transfer protein (MTP) inhibitor used in the treatment of familial hypercholesterolemia