Learn More
Apexbio Technology LLC Lomustine 13010-47-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062351
Encompass_Preferred
Lomustine (CAS 13010-47-4) is an anticancer small molecule belonging to the nitrosourea class of alkylating agents It exerts its antitumor effects primarily by forming DNA cross-links leading to impaired replication and selective cytotoxicity in rapidly dividing tumor cells Lomustine shows inhibitory activity against various cancer cell lines including breast cancer (ZR-75-1 IC50 25 M) astrocytoma (U87MG IC50 8 8 M) and colon carcinoma (LS174T IC50 13 M) Due to its lipophilic nature lomustine efficiently crosses the blood-brain barrier facilitating its use in research involving central nervous system malignancies and related leukemia models
Catalog No. 50-000-62351 Supplier Apexbio Technology LLC Supplier No. B19635.1
Lomustine (CAS 13010-47-4) is an anticancer small molecule belonging to the nitrosourea class of alkylating agents It exerts its antitumor effects primarily by forming DNA cross-links leading to impaired replication and selective cytotoxicity in rapidly dividing tumor cells Lomustine shows inhibitory activity against various cancer cell lines including breast cancer (ZR-75-1 IC50 25 M) astrocytoma (U87MG IC50 8 8 M) and colon carcinoma (LS174T IC50 13 M) Due to its lipophilic nature lomustine efficiently crosses the blood-brain barrier facilitating its use in research involving central nervous system malignancies and related leukemia models