Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Lomustine

Catalog No. AC465490010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465490010 1 g
AC465492500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465490010 Supplier Thermo Scientific Chemicals Supplier No. 465490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 13010-47-4 | C9H16ClN3O2 | 233.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13010-47-4
Molecular Formula C9H16ClN3O2
Molecular Weight (g/mol) 233.70
MDL Number MFCD00012392
InChI Key GQYIWUVLTXOXAJ-UHFFFAOYSA-N
IUPAC Name 3-(2-chloroethyl)-1-cyclohexyl-3-nitrosourea
SMILES ClCCN(N=O)C(=O)NC1CCCCC1

Specifications

Alpha Vector LOMUSTINE
Melting Point 89°C to 93°C
Color Yellow
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 1 g
UN Number 2811
Sensitivity Light sensitive
Solubility Information soluble aq. soln., soluble in acetone, chloroform, ethanol
Formula Weight 233.7
Physical Form Powder
Chemical Name or Material Lomustine
Show More Show Less
Hazard Category GHS06: Toxic
Hazard Statement GHS08: Health hazard
ShelfLife 5 years
storageNote1 Freezer -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.