Learn More
Enzo Life Sciences Lomustine (50mg). CAS: 13010-47-4
SDP
Supplier:  Enzo Life Sciences ALX400038M050
Encompass_Preferred
Possesses antitumor activity similar to carmustine (ALX-400-037). Alternative name: CCNU, N-(2-Chloroethyl)-N\'-cyclohexyl-N-nitrosourea. Purity: ≥97% (HPLC). Appearance: Yellowish crystalline solid. Solubility: Soluble in 100% ethanol or acetone. Long Term Storage: +4°C.
Catalog No. 50-200-9080