Learn More
TARGETMOL CHEMICALS INC LONAFARNIB 10MG
SDP
Catalog No. 502579448
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lonafarnib (Sch66336) is an orally bioavailable FPTase inhibitor for H-ras K-ras-4B and N-ras (IC50 1.9/5.2/2.8 nM). purity: 98%
Catalog No. 50-257-9448 Supplier TARGETMOL CHEMICALS INC Supplier No. T630210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Lonafarnib (Sch66336) is an orally bioavailable FPTase inhibitor for H-ras K-ras-4B and N-ras (IC50 1.9/5.2/2.8 nM). purity: 98%