Learn More
Apexbio Technology LLC Lonafarnib 193275-84-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061627
Encompass_Preferred
Lonafarnib (CAS 193275-84-2) also known as SCH66336 or Sarasar is a tricyclic orally bioavailable inhibitor of farnesyltransferase (FTase) functioning without peptide or sulfhydryl groups FTase mediates post-translational farnesylation of Ras proteins a modification required for their membrane localization and activation of downstream signaling pathways involved in cellular growth proliferation and survival By inhibiting this enzymatic activity Lonafarnib disrupts Ras translocation to the plasma membrane limiting Ras-mediated signaling Currently Lonafarnib is widely evaluated in preclinical and clinical investigations particularly in oncology contexts
Catalog No. 50-000-61627 Supplier Apexbio Technology LLC Supplier No. A43795.1
Lonafarnib (CAS 193275-84-2) also known as SCH66336 or Sarasar is a tricyclic orally bioavailable inhibitor of farnesyltransferase (FTase) functioning without peptide or sulfhydryl groups FTase mediates post-translational farnesylation of Ras proteins a modification required for their membrane localization and activation of downstream signaling pathways involved in cellular growth proliferation and survival By inhibiting this enzymatic activity Lonafarnib disrupts Ras translocation to the plasma membrane limiting Ras-mediated signaling Currently Lonafarnib is widely evaluated in preclinical and clinical investigations particularly in oncology contexts