Learn More
Selleck Chemical LLC Lonafarnib 5mg 193275-84-2 SCH66336
SDP
Supplier:  Selleck Chemical LLC S27975MG
Encompass_Preferred
Lonafarnib (SCH66336) is an orally bioavailable FPTase inhibitor for H-ras, K-ras-4B and N-ras with IC50 of 1.9 nM, 5.2 nM and 2.8 nM in cell-free assays, respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7978