Learn More
Medchemexpress LLC HY-U00156 1mg Medchemexpress, Lonapalene CAS:91431-42-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001561MG
Encompass_Preferred
Medchemexpress, HY-U00156 1mg Lonapalene CAS:91431-42-4 Lonapalene (RS4317) is a topically effective 5-lipoxygenase ( 5-LO ) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4429