Learn More
Medchemexpress LLC Loncastuximab tesirine, (Synonyms: Loncastuximab tesirine-lpyl; ADCT-402), 500ug
SDP
Catalog No. NC2824149
Encompass_Preferred
Loncastuximab tesirine is a human cluster of differentiation 19 (CD19)-directed antibody-drug conjugate (ADC). Once bound to CD19 on the cell membrane, loncastuximab tesirine is rapidly internalised and triggers cell death. Loncastuximab tesirin induces cell apoptosis, it can be used for the research of diffuse large B-cell lymphoma.
Catalog No. NC2824149 Supplier Medchemexpress LLC Supplier No. HYP99349500UG
Loncastuximab tesirine is a human cluster of differentiation 19 (CD19)-directed antibody-drug conjugate (ADC). Once bound to CD19 on the cell membrane, loncastuximab tesirine is rapidly internalised and triggers cell death. Loncastuximab tesirin induces cell apoptosis, it can be used for the research of diffuse large B-cell lymphoma.