Learn More
Electron Microscopy Sciences Maxtaform London Finder Grid, 135 mesh Copper, 188 µm Pitch
SDP
Catalog No. 5026972
Encompass_Preferred
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Copper
Pitch: 188 µm
Catalog No. 50-269-72 Supplier Electron Microscopy Sciences Supplier No. LF135CU
Maxtaform grids with reference patterns are of the highest consistent quality.
Material: Copper
Pitch: 188 µm